Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC6A1
Entrez Gene ID
6529
Gene Name
solute carrier family 6 member 1
Gene Aliases
GABATHG, GABATR, GAT1, MAE
Chromosome Location
Chr.3: 10992718 - 11039249 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003042.3 NP_003033.3 8 - 9 1255 54
XM_005265410.4 XP_005265467.1 8 - 9 2612 54
XM_005265411.4 XP_005265468.1 8 - 9 2551 54
XM_006713306.3 XP_006713369.1 8 - 9 1210 54
XM_011534025.2 XP_011532327.1 8 - 9 3302 54
XM_011534027.2 XP_011532329.1 8 - 9 3241 54
XM_017007071.1 XP_016862560.1 9 - 10 3034 54
XM_017007072.1 XP_016862561.1 9 - 10 2344 54
GenBank mRNA AK295702.1 - 5 - 6 618 54
BC033904.1 - 8 - 9 1223 54
BC057291.1 - 8 - 9 1235 54
X54673.1 - 7 - 8 1078 54

View Full Product Details