Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
Chromosome Location
Chr.16: 68022944 - 68079323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271762.1 NP_001258691.1 10 - 11 1062 67
NM_001271763.1 NP_001258692.1 9 - 10 1037 67
NM_017803.4 NP_060273.1 11 - 12 1142 67
GenBank mRNA AB101210.1 - 11 - 12 806 67
AK000406.1 - 10 - 11 709 67
AK290578.1 - 11 - 12 844 67
AK296663.1 - 9 - 10 689 67
BC006527.2 - 11 - 12 785 67

View Full Product Details