Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
-
Chromosome Location
Chr.1: 240013776 - 240475189 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305424.1 NP_001292353.1 1 - 2 1838 59
NM_020066.4 NP_064450.3 1 - 2 1838 59
XM_011544237.2 XP_011542539.1 1 - 2 1836 59
XM_017001837.1 XP_016857326.1 1 - 2 1836 59
XM_017001838.1 XP_016857327.1 1 - 2 1836 59
XM_017001842.1 XP_016857331.1 1 - 2 1836 59
XM_017001843.1 XP_016857332.1 1 - 2 1836 59
GenBank mRNA AB209153.1 - 1 - 2 431 59
AF218941.1 - 1 - 2 428 59

View Full Product Details