The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006059.3 | NP_006050.3 | 16 - 17 | 2987 | 95 |
XM_006716921.2 | XP_006716984.1 | 16 - 17 | 2930 | 95 | |
XM_011518121.1 | XP_011516423.1 | 16 - 17 | 2930 | 95 | |
GenBank mRNA | AB208887.1 | - | 15 - 16 | 2303 | 95 |
AL551493.3 | - | 4 - 5 | 656 | 95 |