The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006059.3 | NP_006050.3 | 2 - 3 | 776 | 80 |
XM_006716921.2 | XP_006716984.1 | 2 - 3 | 719 | 80 | |
XM_011518121.1 | XP_011516423.1 | 2 - 3 | 719 | 80 | |
GenBank mRNA | AB208887.1 | - | 1 - 2 | 92 | 80 |
AF041835.1 | - | 2 - 3 | 775 | 80 | |
AK075536.1 | - | 2 - 3 | 719 | 80 |