Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002375.4 NP_002366.2 17 - 18 3788 119
XM_005265133.4 XP_005265190.1 20 - 21 6854 119
XM_005265134.4 XP_005265191.1 20 - 21 7267 119
XM_005265155.4 XP_005265212.1 16 - 17 3712 119
XM_005265157.4 XP_005265214.1 10 - 11 3081 119
XM_005265158.2 XP_005265215.1 8 - 9 2874 119
XM_006713146.3 XP_006713209.1 21 - 22 6886 119
XM_006713147.3 XP_006713210.1 21 - 22 7303 119
XM_006713152.3 XP_006713215.1 20 - 21 7252 119
XM_011533703.2 XP_011532005.1 20 - 21 6803 119
XM_011533704.2 XP_011532006.1 20 - 21 6758 119
XM_011533705.1 XP_011532007.1 19 - 20 6752 119
XM_011533708.2 XP_011532010.1 19 - 20 6707 119
XM_011533709.2 XP_011532011.1 19 - 20 6689 119
XM_011533710.2 XP_011532012.1 19 - 20 6680 119
XM_011533718.1 XP_011532020.1 17 - 18 6545 119
XM_011533719.2 XP_011532021.1 18 - 19 3368 119
XM_017006391.1 XP_016861880.1 20 - 21 6758 119
XM_017006392.1 XP_016861881.1 19 - 20 6689 119
XM_017006393.1 XP_016861882.1 20 - 21 6835 119
XM_017006394.1 XP_016861883.1 19 - 20 7129 119
XM_017006395.1 XP_016861884.1 18 - 19 3368 119
XM_017006396.1 XP_016861885.1 18 - 19 3323 119
XM_017006397.1 XP_016861886.1 18 - 19 3323 119
XM_017006398.1 XP_016861887.1 17 - 18 3317 119
XM_017006399.1 XP_016861888.1 18 - 19 3832 119
XM_017006400.1 XP_016861889.1 18 - 19 3832 119
XM_017006401.1 XP_016861890.1 16 - 17 3209 119
XM_017006402.1 XP_016861891.1 17 - 18 3781 119
XM_017006403.1 XP_016861892.1 16 - 17 3161 119
XM_017006404.1 XP_016861893.1 16 - 17 3161 119
XM_017006405.1 XP_016861894.1 17 - 18 3736 119
XM_017006406.1 XP_016861895.1 17 - 18 3718 119
XM_017006407.1 XP_016861896.1 16 - 17 3116 119
XM_017006408.1 XP_016861897.1 16 - 17 3116 119
XM_017006409.1 XP_016861898.1 16 - 17 3667 119
XM_017006410.1 XP_016861899.1 17 - 18 3664 119
XM_017006411.1 XP_016861900.1 16 - 17 3658 119
XM_017006412.1 XP_016861901.1 16 - 17 3658 119
XM_017006413.1 XP_016861902.1 15 - 16 3574 119
XM_017006414.1 XP_016861903.1 15 - 16 3574 119
XM_017006415.1 XP_016861904.1 15 - 16 3529 119
XM_017006416.1 XP_016861905.1 13 - 14 2951 119
XM_017006417.1 XP_016861906.1 10 - 11 3081 119
XM_017006418.1 XP_016861907.1 10 - 11 3036 119
XM_017006419.1 XP_016861908.1 9 - 10 2967 119
XM_017006420.1 XP_016861909.1 9 - 10 2967 119
XM_017006421.1 XP_016861910.1 8 - 9 2874 119
XM_017006422.1 XP_016861911.1 8 - 9 2829 119
GenBank mRNA AL831878.1 - 4 - 5 367 119
BC008715.2 - 15 - 16 2833 119
BC012794.2 - 15 - 16 2833 119
BC015149.2 - 15 - 16 2833 119
BC048200.1 - 10 - 11 2379 119
M64571.1 - 17 - 18 3352 119
U19727.1 - 17 - 18 3337 119

View Full Product Details