Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134364.1 NP_001127836.1 4 - 5 939 76
NM_002375.4 NP_002366.2 4 - 5 939 76
XM_005265133.4 XP_005265190.1 5 - 6 570 76
XM_005265134.4 XP_005265191.1 5 - 6 983 76
XM_005265155.4 XP_005265212.1 3 - 4 863 76
XM_006713146.3 XP_006713209.1 6 - 7 602 76
XM_006713147.3 XP_006713210.1 6 - 7 1019 76
XM_006713152.3 XP_006713215.1 5 - 6 968 76
XM_011533703.2 XP_011532005.1 5 - 6 519 76
XM_011533704.2 XP_011532006.1 5 - 6 519 76
XM_011533705.1 XP_011532007.1 5 - 6 519 76
XM_011533708.2 XP_011532010.1 5 - 6 519 76
XM_011533709.2 XP_011532011.1 5 - 6 519 76
XM_011533710.2 XP_011532012.1 5 - 6 519 76
XM_011533718.1 XP_011532020.1 5 - 6 519 76
XM_011533719.2 XP_011532021.1 5 - 6 519 76
XM_017006391.1 XP_016861880.1 5 - 6 519 76
XM_017006392.1 XP_016861881.1 5 - 6 519 76
XM_017006393.1 XP_016861882.1 5 - 6 551 76
XM_017006394.1 XP_016861883.1 5 - 6 968 76
XM_017006395.1 XP_016861884.1 5 - 6 519 76
XM_017006396.1 XP_016861885.1 5 - 6 519 76
XM_017006397.1 XP_016861886.1 5 - 6 519 76
XM_017006398.1 XP_016861887.1 4 - 5 468 76
XM_017006399.1 XP_016861888.1 5 - 6 983 76
XM_017006400.1 XP_016861889.1 5 - 6 983 76
XM_017006401.1 XP_016861890.1 4 - 5 483 76
XM_017006402.1 XP_016861891.1 4 - 5 932 76
XM_017006403.1 XP_016861892.1 5 - 6 519 76
XM_017006404.1 XP_016861893.1 5 - 6 519 76
XM_017006405.1 XP_016861894.1 4 - 5 932 76
XM_017006406.1 XP_016861895.1 5 - 6 983 76
XM_017006407.1 XP_016861896.1 5 - 6 519 76
XM_017006408.1 XP_016861897.1 5 - 6 519 76
XM_017006409.1 XP_016861898.1 4 - 5 932 76
XM_017006410.1 XP_016861899.1 5 - 6 983 76
XM_017006411.1 XP_016861900.1 4 - 5 932 76
XM_017006412.1 XP_016861901.1 4 - 5 932 76
XM_017006413.1 XP_016861902.1 4 - 5 932 76
XM_017006414.1 XP_016861903.1 4 - 5 932 76
XM_017006415.1 XP_016861904.1 4 - 5 932 76
XM_017006416.1 XP_016861905.1 3 - 4 432 76
GenBank mRNA AB209377.1 - 4 - 5 499 76
AK054577.1 - 4 - 5 934 76
AK297242.1 - 3 - 4 438 76
AK307606.1 - 5 - 6 552 76
BC008715.2 - 4 - 5 503 76
BC012794.2 - 4 - 5 503 76
BC015149.2 - 4 - 5 503 76
BC019229.1 - 4 - 5 469 76
BC067228.1 - 5 - 6 474 76
M64571.1 - 4 - 5 503 76
U19727.1 - 4 - 5 488 76

View Full Product Details