The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171904.1 | NP_001165375.1 | 15 - 16 | 3572 | 65 |
NM_014703.2 | NP_055518.1 | 16 - 17 | 3622 | 65 | |
XM_005276753.4 | XP_005276810.1 | 14 - 15 | 3453 | 65 | |
XM_005276755.4 | XP_005276812.1 | 15 - 16 | 3534 | 65 | |
XM_011534273.2 | XP_011532575.1 | 17 - 18 | 3713 | 65 | |
XM_011534274.2 | XP_011532576.1 | 17 - 18 | 3773 | 65 | |
XM_011534275.2 | XP_011532577.1 | 17 - 18 | 3688 | 65 | |
XM_011534276.2 | XP_011532578.1 | 17 - 18 | 3735 | 65 | |
XM_011534277.2 | XP_011532579.1 | 15 - 16 | 3606 | 65 | |
XM_017007546.1 | XP_016863035.1 | 16 - 17 | 3631 | 65 | |
XM_017007547.1 | XP_016863036.1 | 16 - 17 | 3588 | 65 | |
XM_017007548.1 | XP_016863037.1 | 16 - 17 | 3723 | 65 | |
XM_017007549.1 | XP_016863038.1 | 17 - 18 | 3773 | 65 | |
XM_017007550.1 | XP_016863039.1 | 17 - 18 | 3773 | 65 | |
XM_017007551.1 | XP_016863040.1 | 14 - 15 | 3483 | 65 | |
GenBank mRNA | AB018343.1 | - | 16 - 17 | 3605 | 65 |
AF061935.1 | - | 11 - 12 | 3120 | 65 | |
AK023239.1 | - | 2 - 3 | 1417 | 65 | |
AL080145.1 | - | 7 - 8 | 2317 | 65 | |
BC022792.1 | - | 11 - 12 | 2289 | 65 | |
BC110371.1 | - | 15 - 16 | 3562 | 65 |