Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF1
Entrez Gene ID
9730
Gene Name
DDB1 and CUL4 associated factor 1
Gene Aliases
VPRBP
Chromosome Location
Chr.3: 51395867 - 51505620 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171904.1 NP_001165375.1 16 - 17 3646 98
NM_014703.2 NP_055518.1 17 - 18 3696 98
XM_005276753.4 XP_005276810.1 15 - 16 3527 98
XM_005276755.4 XP_005276812.1 16 - 17 3608 98
XM_011534273.2 XP_011532575.1 18 - 19 3787 98
XM_011534274.2 XP_011532576.1 18 - 19 3847 98
XM_011534275.2 XP_011532577.1 18 - 19 3762 98
XM_011534276.2 XP_011532578.1 18 - 19 3809 98
XM_011534277.2 XP_011532579.1 16 - 17 3680 98
XM_017007546.1 XP_016863035.1 17 - 18 3705 98
XM_017007547.1 XP_016863036.1 17 - 18 3662 98
XM_017007548.1 XP_016863037.1 17 - 18 3797 98
XM_017007549.1 XP_016863038.1 18 - 19 3847 98
XM_017007550.1 XP_016863039.1 18 - 19 3847 98
XM_017007551.1 XP_016863040.1 15 - 16 3557 98
GenBank mRNA AB018343.1 - 17 - 18 3679 98
AF061935.1 - 12 - 13 3194 98
AK023239.1 - 3 - 4 1491 98
AL080145.1 - 8 - 9 2391 98
BC022792.1 - 12 - 13 2363 98
BC110371.1 - 16 - 17 3636 98

View Full Product Details