Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
Chromosome Location
Chr.3: 49108046 - 49120938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199160.1 NP_001186089.1 7 - 8 1449 61
NM_001199161.1 NP_001186090.1 7 - 8 1449 61
NM_001199162.1 NP_001186091.1 7 - 8 1419 61
NM_006677.2 NP_006668.1 6 - 7 1146 61
XM_005264825.2 XP_005264882.1 7 - 8 1444 61
XM_005264831.2 XP_005264888.1 6 - 7 1096 61
XM_006712947.2 XP_006713010.1 7 - 8 1396 61
XM_006712948.2 XP_006713011.1 7 - 8 1444 61
XM_006712950.2 XP_006713013.1 7 - 8 1399 61
XM_006712951.2 XP_006713014.1 7 - 8 1396 61
XM_006712952.2 XP_006713015.1 7 - 8 1399 61
XM_006712953.1 XP_006713016.1 7 - 8 1370 61
XM_017005615.1 XP_016861104.1 7 - 8 1441 61
XM_017005616.1 XP_016861105.1 7 - 8 1441 61
XM_017005617.1 XP_016861106.1 7 - 8 1399 61
XM_017005619.1 XP_016861108.1 7 - 8 1441 61
XM_017005620.1 XP_016861109.1 7 - 8 1441 61
XM_017005621.1 XP_016861110.1 7 - 8 1294 61
XM_017005623.1 XP_016861112.1 7 - 8 1294 61
XM_017005624.1 XP_016861113.1 7 - 8 1294 61
XM_017005625.1 XP_016861114.1 6 - 7 1141 61
XM_017005626.1 XP_016861115.1 6 - 7 1160 61
XM_017005628.1 XP_016861117.1 6 - 7 1096 61
XM_017005629.1 XP_016861118.1 6 - 7 1217 61
XM_017005631.1 XP_016861120.1 6 - 7 1141 61
XM_017005633.1 XP_016861122.1 6 - 7 1141 61
GenBank mRNA AB020698.1 - 6 - 7 988 61
AK294756.1 - 7 - 8 1342 61
AK296447.1 - 7 - 8 1284 61
AK300425.1 - 7 - 8 1273 61
BC014934.1 - 2 - 3 421 61
BC106029.1 - 7 - 8 1267 61
BC142660.1 - 7 - 8 1319 61
BC142727.1 - 7 - 8 1325 61
BC146752.1 - 6 - 7 988 61

View Full Product Details