Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBMS3
Entrez Gene ID
27303
Gene Name
RNA binding motif single stranded interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.3: 29280860 - 30010395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003792.2 NP_001003792.1 2 - 3 607 102
NM_001003793.2 NP_001003793.1 2 - 3 610 102
NM_001177711.1 NP_001171182.1 2 - 3 610 102
NM_001177712.1 NP_001171183.1 2 - 3 610 102
NM_014483.3 NP_055298.2 2 - 3 610 102
XM_005265060.2 XP_005265117.1 2 - 3 1059 102
XM_005265061.2 XP_005265118.1 2 - 3 1062 102
XM_005265062.1 XP_005265119.1 2 - 3 1062 102
XM_017006178.1 XP_016861667.1 2 - 3 1059 102
XM_017006179.1 XP_016861668.1 2 - 3 1059 102
XM_017006180.1 XP_016861669.1 2 - 3 1062 102
XM_017006181.1 XP_016861670.1 2 - 3 1059 102
XM_017006182.1 XP_016861671.1 3 - 4 1138 102
GenBank mRNA AF023259.1 - 2 - 3 387 102
AK292789.1 - 2 - 3 286 102
AY236871.1 - 2 - 3 470 102
AY236872.1 - 2 - 3 467 102
BC117315.1 - 2 - 3 307 102
BC143521.1 - 2 - 3 307 102

View Full Product Details