The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003792.2 | NP_001003792.1 | 3 - 4 | 666 | 105 |
NM_001003793.2 | NP_001003793.1 | 3 - 4 | 669 | 105 | |
NM_001177711.1 | NP_001171182.1 | 3 - 4 | 669 | 105 | |
NM_001177712.1 | NP_001171183.1 | 3 - 4 | 669 | 105 | |
NM_014483.3 | NP_055298.2 | 3 - 4 | 669 | 105 | |
XM_005265060.2 | XP_005265117.1 | 3 - 4 | 1118 | 105 | |
XM_005265061.2 | XP_005265118.1 | 3 - 4 | 1121 | 105 | |
XM_005265062.1 | XP_005265119.1 | 3 - 4 | 1121 | 105 | |
XM_017006178.1 | XP_016861667.1 | 3 - 4 | 1118 | 105 | |
XM_017006179.1 | XP_016861668.1 | 3 - 4 | 1118 | 105 | |
XM_017006180.1 | XP_016861669.1 | 3 - 4 | 1121 | 105 | |
XM_017006181.1 | XP_016861670.1 | 3 - 4 | 1118 | 105 | |
XM_017006182.1 | XP_016861671.1 | 4 - 5 | 1197 | 105 | |
GenBank mRNA | AF023259.1 | - | 3 - 4 | 446 | 105 |
AK292789.1 | - | 3 - 4 | 345 | 105 | |
AY236871.1 | - | 3 - 4 | 529 | 105 | |
AY236872.1 | - | 3 - 4 | 526 | 105 | |
BC117315.1 | - | 3 - 4 | 366 | 105 | |
BC143521.1 | - | 3 - 4 | 366 | 105 |