The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284413.1 | NP_001271342.1 | 15 - 16 | 2014 | 122 |
NM_001284415.1 | NP_001271344.1 | 14 - 15 | 1915 | 122 | |
NM_001284416.1 | NP_001271345.1 | 13 - 14 | 1849 | 122 | |
NM_012260.3 | NP_036392.2 | 16 - 17 | 2095 | 122 | |
NR_104315.1 | 14 - 15 | 1916 | 122 | ||
GenBank mRNA | AA643224.1 | - | 1 - 2 | 122 | |
AF161397.1 | - | 16 - 17 | 1715 | 122 | |
AJ131753.1 | - | 16 - 17 | 1805 | 122 | |
AK056977.1 | - | 8 - 9 | 1089 | 122 | |
AK301546.1 | - | 15 - 16 | 1731 | 122 | |
AK301990.1 | - | 13 - 14 | 1584 | 122 | |
AK302086.1 | - | 14 - 15 | 1632 | 122 | |
BC001627.1 | - | 16 - 17 | 1813 | 122 | |
BC007440.2 | - | 14 - 15 | 1621 | 122 |