The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284413.1 | NP_001271342.1 | 2 - 3 | 574 | 68 |
NM_001284415.1 | NP_001271344.1 | 2 - 3 | 574 | 68 | |
NM_001284416.1 | NP_001271345.1 | 2 - 3 | 574 | 68 | |
NM_012260.3 | NP_036392.2 | 2 - 3 | 574 | 68 | |
NR_104315.1 | 2 - 3 | 574 | 68 | ||
GenBank mRNA | AF161397.1 | - | 2 - 3 | 194 | 68 |
AJ131753.1 | - | 2 - 3 | 284 | 68 | |
AK301546.1 | - | 2 - 3 | 291 | 68 | |
AK301990.1 | - | 2 - 3 | 309 | 68 | |
AK302086.1 | - | 2 - 3 | 291 | 68 | |
BC001627.1 | - | 2 - 3 | 292 | 68 | |
BC007440.2 | - | 2 - 3 | 279 | 68 | |
DA609383.1 | - | 2 - 3 | 291 | 68 | |
DB049608.1 | - | 2 - 3 | 345 | 68 |