The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007193.4 | NP_009124.2 | 5 - 6 | 563 | 118 |
XM_011531571.2 | XP_011529873.1 | 6 - 7 | 635 | 118 | |
GenBank mRNA | AF196478.1 | - | 5 - 6 | 525 | 118 |
AJ238979.1 | - | 5 - 6 | 516 | 118 | |
BC007320.2 | - | 5 - 6 | 543 | 118 |