The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012464.4 | NP_036596.3 | 11 - 12 | 2025 | 100 |
XM_011532214.1 | XP_011530516.1 | 11 - 12 | 2083 | 100 | |
XM_017008570.1 | XP_016864059.1 | 12 - 13 | 2151 | 100 | |
GenBank mRNA | AF282732.1 | - | 11 - 12 | 1629 | 100 |
BC136429.1 | - | 11 - 12 | 1935 | 100 | |
BC136430.1 | - | 11 - 12 | 1935 | 100 | |
BC144085.1 | - | 11 - 12 | 1935 | 100 | |
U91963.1 | - | 11 - 12 | 2025 | 100 |