Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FRMD4A
- Entrez Gene ID
-
55691
- Gene Name
- FERM domain containing 4A
- Gene Aliases
-
CCAFCA, FRMD4, bA295P9.4
- Chromosome Location
-
Chr.10: 13643706 - 14330952 on Build GRCh38
View Full Product Details