Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK3
Entrez Gene ID
354
Gene Name
kallikrein related peptidase 3
Gene Aliases
APS, KLK2A1, PSA, hK3
Chromosome Location
Chr.19: 50854915 - 50860764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030048.1 NP_001025219.1 2 - 3 253 65
GenBank mRNA BQ932072.1 - 2 - 3 219 65

View Full Product Details