The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019112.3 | NP_061985.2 | 5 - 6 | 650 | 77 |
XM_006722616.1 | XP_006722679.1 | 5 - 6 | 637 | 77 | |
XM_006722617.2 | XP_006722680.1 | 5 - 6 | 637 | 77 | |
XM_011527628.2 | XP_011525930.1 | 5 - 6 | 511 | 77 | |
XM_011527629.1 | XP_011525931.1 | 5 - 6 | 637 | 77 | |
XM_011527630.1 | XP_011525932.1 | 5 - 6 | 637 | 77 | |
XM_011527631.1 | XP_011525933.1 | 5 - 6 | 637 | 77 | |
XM_011527633.2 | XP_011525935.1 | 5 - 6 | 637 | 77 | |
XM_011527634.1 | XP_011525936.1 | 5 - 6 | 637 | 77 | |
XM_011527635.1 | XP_011525937.1 | 5 - 6 | 637 | 77 | |
XM_017026142.1 | XP_016881631.1 | 5 - 6 | 637 | 77 | |
XM_017026143.1 | XP_016881632.1 | 5 - 6 | 637 | 77 | |
GenBank mRNA | AF250238.1 | - | 4 - 5 | 419 | 77 |
AF328787.1 | - | 4 - 5 | 535 | 77 | |
AK301108.1 | - | 5 - 6 | 630 | 77 | |
AK310127.1 | - | 5 - 6 | 637 | 77 | |
AK310909.1 | - | 3 - 4 | 944 | 77 |