Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
Chromosome Location
Chr.20: 50586108 - 50692004 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290268.1 NP_001277197.1 6 - 7 835 78
NM_080829.3 NP_543019.2 6 - 7 768 78
NR_110890.1 6 - 7 1020 78
XM_005260294.3 XP_005260351.1 7 - 8 832 78
XM_006723713.3 XP_006723776.1 6 - 7 880 78
XM_011528578.2 XP_011526880.1 7 - 8 860 78
XM_011528579.2 XP_011526881.1 7 - 8 998 78
XM_011528580.2 XP_011526882.1 7 - 8 1052 78
XM_011528581.2 XP_011526883.1 7 - 8 1036 78
XM_011528584.2 XP_011526886.1 6 - 7 897 78
XM_011528586.2 XP_011526888.1 6 - 7 898 78
XM_017027682.1 XP_016883171.1 6 - 7 897 78
GenBank mRNA AK056792.1 - 6 - 7 739 78
AK295781.1 - 6 - 7 711 78
AK299337.1 - 6 - 7 684 78

View Full Product Details