Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNFX1
Entrez Gene ID
57169
Gene Name
zinc finger NFX1-type containing 1
Gene Aliases
-
Chromosome Location
Chr.20: 49245897 - 49278244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK002139.1 - 1 - 2 110 121

View Full Product Details