The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282806.1 | NP_001269735.1 | 1 - 2 | 144 | 103 |
NM_022826.3 | NP_073737.1 | 1 - 2 | 144 | 103 | |
XM_017004718.1 | XP_016860207.1 | 1 - 2 | 144 | 103 | |
XM_017004720.1 | XP_016860209.1 | 1 - 2 | 132 | 103 | |
GenBank mRNA | AK022973.1 | - | 1 - 2 | 102 | 103 |
AK222880.1 | - | 1 - 2 | 94 | 103 | |
AK299051.1 | - | 1 - 2 | 112 | 103 | |
BC065014.1 | - | 1 - 2 | 68 | 103 |