Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH7
Entrez Gene ID
64844
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
AXO, AXOT, MARCH-VII, RNF177
Chromosome Location
Chr.2: 159712457 - 159768583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282805.1 NP_001269734.1 4 - 5 441 120
NM_001282806.1 NP_001269735.1 2 - 3 313 120
NM_022826.3 NP_073737.1 2 - 3 313 120
XM_005246773.2 XP_005246830.1 2 - 3 336 120
XM_005246774.1 XP_005246831.1 2 - 3 336 120
XM_017004718.1 XP_016860207.1 2 - 3 313 120
XM_017004719.1 XP_016860208.1 2 - 3 339 120
XM_017004720.1 XP_016860209.1 2 - 3 301 120
XM_017004721.1 XP_016860210.1 2 - 3 331 120
GenBank mRNA AK022973.1 - 2 - 3 271 120
AK222880.1 - 2 - 3 263 120
AK292836.1 - 4 - 5 409 120
AK299051.1 - 2 - 3 281 120
BC065014.1 - 2 - 3 237 120
DA825046.1 - 2 - 3 317 120

View Full Product Details