The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017916.2 | NP_060386.1 | 1 - 2 | 376 | 59 |
GenBank mRNA | AK000650.1 | - | 1 - 2 | 328 | 59 |
AK294687.1 | - | 1 - 2 | 302 | 59 | |
BC001108.2 | - | 1 - 2 | 306 | 59 | |
DC385084.1 | - | 1 - 2 | 376 | 59 |