Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPG
Entrez Gene ID
822
Gene Name
capping actin protein, gelsolin like
Gene Aliases
AFCP, HEL-S-66, MCP
Chromosome Location
Chr.2: 85394748 - 85418467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256139.1 NP_001243068.1 6 - 7 772 110
NM_001320732.1 NP_001307661.1 6 - 7 748 110
NM_001320733.1 NP_001307662.1 6 - 7 709 110
NM_001747.3 NP_001738.2 6 - 7 1062 110
XM_011533122.1 XP_011531424.1 6 - 7 882 110
XM_011533123.1 XP_011531425.1 6 - 7 778 110
GenBank mRNA AK130130.1 - 6 - 7 1062 110
AK300507.1 - 6 - 7 654 110
AK309290.1 - 5 - 6 431 110
AK313898.1 - 6 - 7 716 110
BC000728.2 - 6 - 7 716 110
BC014549.1 - 6 - 7 718 110
BU626913.1 - 4 - 5 110
FJ224312.1 - 6 - 7 918 110
M94345.1 - 6 - 7 717 110

View Full Product Details