The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006634.2 | NP_006625.1 | 2 - 3 | 223 | 68 |
GenBank mRNA | AF054825.1 | - | 2 - 3 | 197 | 68 |
AF077197.1 | - | 1 - 2 | 150 | 68 | |
AF151025.1 | - | 2 - 3 | 181 | 68 | |
AK292660.1 | - | 2 - 3 | 221 | 68 | |
BC017891.1 | - | 2 - 3 | 223 | 68 | |
CR542215.1 | - | 1 - 2 | 140 | 68 |