Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC31
Entrez Gene ID
64427
Gene Name
tetratricopeptide repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.2: 74483032 - 74494564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022492.4 NP_071937.4 3 - 4 250 104
XM_005264492.1 XP_005264549.1 3 - 4 239 104
XM_011533035.1 XP_011531337.1 3 - 4 239 104
XM_011533036.1 XP_011531338.1 3 - 4 239 104
XM_011533037.1 XP_011531339.1 3 - 4 239 104
XM_017004706.1 XP_016860195.1 3 - 4 239 104
XM_017004707.1 XP_016860196.1 3 - 4 239 104
XM_017004709.1 XP_016860198.1 2 - 3 374 104
GenBank mRNA AK022850.1 - 3 - 4 236 104
AK223355.1 - 3 - 4 234 104
AK225626.1 - 3 - 4 236 104
AK303245.1 - 3 - 4 236 104
BC036017.1 - 3 - 4 234 104
BC096709.1 - 3 - 4 236 104
BC098365.1 - 3 - 4 236 104

View Full Product Details