The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005743.1 | NP_001005743.1 | 8 - 9 | 767 | 75 |
NM_001005744.1 | NP_001005744.1 | 8 - 9 | 767 | 75 | |
NM_001005745.1 | NP_001005745.1 | 7 - 8 | 734 | 75 | |
NM_001320114.1 | NP_001307043.1 | 8 - 9 | 787 | 75 | |
NM_003744.5 | NP_003735.3 | 7 - 8 | 734 | 75 | |
GenBank mRNA | AF015040.1 | - | 8 - 9 | 725 | 75 |
AF171938.1 | - | 8 - 9 | 717 | 75 | |
AF171939.1 | - | 8 - 9 | 717 | 75 | |
AF171940.1 | - | 7 - 8 | 684 | 75 | |
AF171941.1 | - | 7 - 8 | 684 | 75 | |
AK023670.1 | - | 2 - 3 | 804 | 75 | |
AK226069.1 | - | 2 - 3 | 2303 | 75 | |
AK304193.1 | - | 6 - 7 | 570 | 75 | |
AK315130.1 | - | 6 - 7 | 615 | 75 | |
BC020788.1 | - | 6 - 7 | 615 | 75 | |
BC068476.1 | - | 7 - 8 | 734 | 75 | |
BX248073.1 | - | 7 - 8 | 624 | 75 | |
BX248775.1 | - | 6 - 7 | 588 | 75 | |
BX248781.1 | - | 6 - 7 | 629 | 75 | |
BX647137.1 | - | 8 - 9 | 726 | 75 | |
EU265734.1 | - | 6 - 7 | 486 | 75 | |
EU265735.1 | - | 5 - 6 | 453 | 75 | |
EU265736.1 | - | 6 - 7 | 486 | 75 | |
EU265737.1 | - | 5 - 6 | 453 | 75 |