The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000578.3 | NP_000569.3 | 10 - 11 | 1382 | 78 |
XM_005246793.3 | XP_005246850.1 | 9 - 10 | 1502 | 78 | |
XM_005246794.3 | XP_005246851.1 | 11 - 12 | 1719 | 78 | |
XM_006712709.3 | XP_006712772.1 | 10 - 11 | 1576 | 78 | |
XM_006712710.3 | XP_006712773.1 | 9 - 10 | 1453 | 78 | |
XM_006712711.3 | XP_006712774.1 | 8 - 9 | 1379 | 78 | |
XM_011511684.2 | XP_011509986.1 | 11 - 12 | 1754 | 78 | |
XM_011511685.2 | XP_011509987.1 | 10 - 11 | 1611 | 78 | |
XM_017004765.1 | XP_016860254.1 | 9 - 10 | 1522 | 78 | |
XM_017004766.1 | XP_016860255.1 | 10 - 11 | 1613 | 78 | |
GenBank mRNA | AB209650.1 | - | 8 - 9 | 3096 | 78 |
AK298667.1 | - | 10 - 11 | 1136 | 78 | |
AK303398.1 | - | 10 - 11 | 1106 | 78 | |
AK307954.1 | - | 10 - 11 | 1322 | 78 | |
AK310914.1 | - | 10 - 11 | 1138 | 78 | |
AK311405.1 | - | 9 - 10 | 989 | 78 | |
BC033754.1 | - | 8 - 9 | 1388 | 78 | |
BC071165.1 | - | 11 - 12 | 1176 | 78 | |
D38171.1 | - | 8 - 9 | 889 | 78 | |
D50402.1 | - | 10 - 11 | 1217 | 78 | |
D50403.1 | - | 10 - 11 | 1128 | 78 | |
L32185.1 | - | 10 - 11 | 1118 | 78 |