The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168465.1 | NP_001161937.1 | 15 - 16 | 2185 | 90 |
NM_001168466.1 | NP_001161938.1 | 13 - 14 | 1927 | 90 | |
NM_001168467.1 | NP_001161939.1 | 14 - 15 | 1899 | 90 | |
NM_152780.3 | NP_689993.2 | 14 - 15 | 2062 | 90 | |
XM_005274478.2 | XP_005274535.1 | 15 - 16 | 2048 | 90 | |
XM_005274479.2 | XP_005274536.1 | 15 - 16 | 2015 | 90 | |
XM_005274480.3 | XP_005274537.1 | 14 - 15 | 1949 | 90 | |
XM_006724481.2 | XP_006724544.1 | 16 - 17 | 2195 | 90 | |
XM_006724482.2 | XP_006724545.1 | 15 - 16 | 2072 | 90 | |
XM_011545483.1 | XP_011543785.1 | 16 - 17 | 2162 | 90 | |
XM_011545484.1 | XP_011543786.1 | 16 - 17 | 2145 | 90 | |
XM_011545485.1 | XP_011543787.1 | 16 - 17 | 2080 | 90 | |
XM_017029393.1 | XP_016884882.1 | 15 - 16 | 2039 | 90 | |
XM_017029394.1 | XP_016884883.1 | 15 - 16 | 2022 | 90 | |
XM_017029395.1 | XP_016884884.1 | 15 - 16 | 1998 | 90 | |
XM_017029396.1 | XP_016884885.1 | 15 - 16 | 1957 | 90 | |
XM_017029397.1 | XP_016884886.1 | 15 - 16 | 1933 | 90 | |
GenBank mRNA | AK027409.1 | - | 14 - 15 | 1964 | 90 |
AK127056.1 | - | 12 - 13 | 1601 | 90 | |
AK294788.1 | - | 14 - 15 | 1893 | 90 | |
AK296319.1 | - | 13 - 14 | 1838 | 90 | |
AK303019.1 | - | 9 - 10 | 1173 | 90 | |
AK316117.1 | - | 13 - 14 | 1835 | 90 | |
BC037165.2 | - | 12 - 13 | 1729 | 90 | |
BC089400.1 | - | 15 - 16 | 2106 | 90 | |
BC136379.1 | - | 14 - 15 | 1979 | 90 | |
KP849473.1 | - | 15 - 16 | 2020 | 90 | |
KP849474.1 | - | 15 - 16 | 1987 | 90 | |
KP979782.1 | - | 15 - 16 | 2054 | 90 | |
KP979783.1 | - | 15 - 16 | 2021 | 90 | |
KP979784.1 | - | 15 - 16 | 2078 | 90 | |
KP979785.1 | - | 15 - 16 | 2045 | 90 | |
KP979786.1 | - | 15 - 16 | 2078 | 90 |