The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003188.3 | NP_003179.1 | 6 - 7 | 1022 | 106 |
NM_145331.2 | NP_663304.1 | 6 - 7 | 1022 | 106 | |
NM_145332.2 | NP_663305.1 | 6 - 7 | 1022 | 106 | |
NM_145333.2 | NP_663306.1 | 6 - 7 | 1022 | 106 | |
XM_006715553.2 | XP_006715616.1 | 6 - 7 | 810 | 106 | |
XM_017011226.1 | XP_016866715.1 | 6 - 7 | 810 | 106 | |
GenBank mRNA | AB009356.1 | - | 6 - 7 | 766 | 106 |
AB009357.1 | - | 6 - 7 | 766 | 106 | |
AB009358.2 | - | 6 - 7 | 604 | 106 | |
AF218074.1 | - | 6 - 7 | 610 | 106 | |
AK299268.1 | - | 4 - 5 | 570 | 106 | |
AK315774.1 | - | 6 - 7 | 790 | 106 | |
BC017715.2 | - | 6 - 7 | 789 | 106 | |
BT019654.1 | - | 6 - 7 | 604 | 106 | |
BT019655.1 | - | 6 - 7 | 604 | 106 |