The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130144.2 | NP_001123616.1 | 18 - 19 | 2860 | 86 |
NM_001164266.1 | NP_001157738.1 | 18 - 19 | 2856 | 86 | |
NM_021070.4 | NP_066548.2 | 18 - 19 | 2860 | 86 | |
XM_011545032.2 | XP_011543334.1 | 18 - 19 | 3042 | 86 | |
XM_011545033.2 | XP_011543335.1 | 18 - 19 | 3041 | 86 | |
GenBank mRNA | AF135960.3 | - | 18 - 19 | 2591 | 86 |
AF318354.1 | - | 16 - 17 | 1892 | 86 | |
AK024477.1 | - | 18 - 19 | 2857 | 86 | |
AK090750.1 | - | 3 - 4 | 497 | 86 | |
AK097212.1 | - | 3 - 4 | 806 | 86 | |
AK124524.1 | - | 7 - 8 | 1924 | 86 | |
AK298858.1 | - | 13 - 14 | 1542 | 86 | |
AK299351.1 | - | 19 - 20 | 2522 | 86 | |
AL117551.1 | - | 2 - 3 | 215 | 86 | |
BC008761.2 | - | 9 - 10 | 1053 | 86 | |
BC136277.1 | - | 18 - 19 | 2666 | 86 |