The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014834.2 | NP_001014834.1 | 2 - 3 | 369 | 75 |
NM_001014835.1 | NP_001014835.1 | 3 - 4 | 442 | 75 | |
XM_011526320.1 | XP_011524622.1 | 3 - 4 | 432 | 75 | |
GenBank mRNA | AB032968.1 | - | 3 - 4 | 388 | 75 |
AK294586.1 | - | 2 - 3 | 372 | 75 | |
AK304651.1 | - | 2 - 3 | 349 | 75 | |
AL834236.1 | - | 2 - 3 | 343 | 75 | |
AX775863.1 | - | 2 - 3 | 160 | 75 | |
AX775865.1 | - | 2 - 3 | 160 | 75 | |
AX775867.1 | - | 2 - 3 | 343 | 75 |