The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282189.1 | NP_001269118.1 | 5 - 6 | 439 | 98 |
NM_001282199.1 | NP_001269128.1 | 4 - 5 | 427 | 98 | |
NM_005671.3 | NP_005662.1 | 5 - 6 | 550 | 98 | |
XM_011544655.2 | XP_011542957.1 | 7 - 8 | 1627 | 98 | |
XM_017013862.1 | XP_016869351.1 | 6 - 7 | 739 | 98 | |
GenBank mRNA | AK296600.1 | - | 5 - 6 | 724 | 98 |
AY302140.1 | - | 5 - 6 | 414 | 98 | |
BC020694.1 | - | 5 - 6 | 534 | 98 | |
BI462115.1 | - | 5 - 6 | 555 | 98 | |
BI755534.1 | - | 4 - 5 | 403 | 98 | |
D83767.1 | - | 5 - 6 | 540 | 98 | |
D83768.1 | - | 3 - 4 | 277 | 98 |