Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr.4: 128809623 - 128875224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287439.1 NP_001274368.1 4 - 5 697 85
NM_001287440.1 NP_001274369.1 4 - 5 494 85
NM_001287441.1 NP_001274370.1 4 - 5 458 85
NM_001287442.1 NP_001274371.1 4 - 5 451 85
NM_001287443.1 NP_001274372.1 4 - 5 455 85
NM_024900.4 NP_079176.2 4 - 5 579 85
NM_199320.3 NP_955352.1 4 - 5 579 85
XM_005263232.1 XP_005263289.1 4 - 5 421 85
XM_017008626.1 XP_016864115.1 4 - 5 536 85
XM_017008627.1 XP_016864116.1 4 - 5 583 85
GenBank mRNA AF520952.1 - 4 - 5 476 85
AK027620.1 - 4 - 5 499 85
BC032376.1 - 4 - 5 413 85
DA478923.1 - 4 - 5 493 85
DA714593.1 - 4 - 5 451 85
DB085413.1 - 4 - 5 455 85
DB293142.1 - 4 - 5 433 85

View Full Product Details