The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145056.1 | NP_001138528.1 | 17 - 18 | 1773 | 82 |
NM_020428.3 | NP_065161.3 | 17 - 18 | 1832 | 82 | |
XM_005259997.1 | XP_005260054.1 | 17 - 18 | 1832 | 82 | |
XM_005259999.1 | XP_005260056.1 | 17 - 18 | 1773 | 82 | |
GenBank mRNA | AK027519.1 | - | 17 - 18 | 1717 | 82 |
AK075512.1 | - | 6 - 7 | 734 | 82 | |
AK126550.1 | - | 5 - 6 | 523 | 82 | |
AK314582.1 | - | 17 - 18 | 1774 | 82 | |
AL832517.1 | - | 7 - 8 | 878 | 82 | |
AL832978.1 | - | 14 - 15 | 1516 | 82 | |
BC010617.2 | - | 14 - 15 | 1523 | 82 | |
BC040556.1 | - | 17 - 18 | 1774 | 82 | |
BC042692.1 | - | 17 - 18 | 1774 | 82 | |
FN668568.1 | - | 17 - 18 | 1832 | 82 |