The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033109.4 | NP_149100.2 | 14 - 15 | 1330 | 117 |
XM_005264629.2 | XP_005264686.1 | 14 - 15 | 1260 | 117 | |
XM_017005172.1 | XP_016860661.1 | 13 - 14 | 1210 | 117 | |
GenBank mRNA | AJ458465.1 | - | 14 - 15 | 1261 | 117 |
AK307589.1 | - | - | 1260 | 117 | |
AY027528.1 | - | 14 - 15 | 1257 | 117 | |
BC053660.1 | - | 14 - 15 | 1244 | 117 |