The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033109.4 | NP_149100.2 | 3 - 4 | 382 | 67 |
XM_005264629.2 | XP_005264686.1 | 3 - 4 | 312 | 67 | |
XM_017005172.1 | XP_016860661.1 | 2 - 3 | 262 | 67 | |
GenBank mRNA | AJ458465.1 | - | 3 - 4 | 313 | 67 |
AK307589.1 | - | - | 312 | 67 | |
AY027528.1 | - | 3 - 4 | 309 | 67 | |
BC053660.1 | - | 3 - 4 | 296 | 67 | |
DA998523.1 | - | 3 - 4 | 348 | 67 | |
DB461021.1 | - | 3 - 4 | 382 | 67 |