The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007008.2 | NP_008939.1 | 1 - 2 | 243 | 87 |
GenBank mRNA | AF077050.1 | - | 1 - 2 | 242 | 87 |
AF087901.1 | - | 1 - 2 | 134 | 87 | |
AF125103.1 | - | 1 - 2 | 243 | 87 | |
AF132048.1 | - | 1 - 2 | 149 | 87 | |
AF177332.1 | - | 1 - 2 | 148 | 87 | |
AY102276.1 | - | 1 - 2 | 263 | 87 | |
BC001035.2 | - | 1 - 2 | 214 | 87 | |
BC007109.1 | - | 1 - 2 | 140 | 87 | |
BC014366.1 | - | 1 - 2 | 140 | 87 |