Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGAP1
Entrez Gene ID
57522
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases
ARHGAP13, NMTC2
Chromosome Location
Chr.12: 63844372 - 64147863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020762.2 NP_065813.1 19 - 20 2464 114
XM_005269042.3 XP_005269099.1 19 - 20 2784 114
XM_011538580.1 XP_011536882.1 20 - 21 2572 114
XM_011538581.2 XP_011536883.1 20 - 21 4077 114
GenBank mRNA AB037725.1 - 18 - 19 3686 114
AK023899.1 - 9 - 10 1194 114
BC053903.1 - 19 - 20 2464 114

Gene Symbol
SLC4A1
Entrez Gene ID
6521
Gene Name
solute carrier family 4 member 1 (Diego blood group)
Gene Aliases
AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR
Chromosome Location
Chr.17: 44248390 - 44268161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF113686.1 - - 114

View Full Product Details