Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SRGAP1
- Entrez Gene ID
-
57522
- Gene Name
- SLIT-ROBO Rho GTPase activating protein 1
- Gene Aliases
-
ARHGAP13, NMTC2
- Chromosome Location
-
Chr.12: 63844372 - 64147863 on Build GRCh38
- Gene Symbol
- SLC4A1
- Entrez Gene ID
-
6521
- Gene Name
- solute carrier family 4 member 1 (Diego blood group)
- Gene Aliases
-
AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR
- Chromosome Location
-
Chr.17: 44248390 - 44268161 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF113686.1 |
- |
- |
|
84 |
View Full Product Details