The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005269042.3 | XP_005269099.1 | 10 - 11 | 1854 | 87 |
GenBank mRNA | AB037725.1 | - | 9 - 10 | 2756 | 87 |