The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308379.1 | NP_001295308.1 | 2 - 3 | 370 | 83 |
NM_015896.3 | NP_056980.2 | 2 - 3 | 370 | 83 | |
GenBank mRNA | AK058135.1 | - | 2 - 3 | 300 | 83 |
AK096525.1 | - | 2 - 3 | 1478 | 83 | |
AK223343.1 | - | 2 - 3 | 285 | 83 | |
AK308086.1 | - | 2 - 3 | 269 | 83 | |
AK315123.1 | - | 2 - 3 | 356 | 83 | |
AL833828.1 | - | 2 - 3 | 299 | 83 | |
BC033732.1 | - | 2 - 3 | 266 | 83 | |
DB036846.1 | - | 2 - 3 | 370 | 83 | |
U70824.1 | - | 2 - 3 | 324 | 83 | |
U70880.1 | - | 2 - 3 | 324 | 83 |