Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
Chromosome Location
Chr.3: 50341106 - 50346028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308379.1 NP_001295308.1 8 - 9 1155 90
NM_015896.3 NP_056980.2 9 - 10 1170 90
XM_005265216.2 XP_005265273.1 8 - 9 1343 90
GenBank mRNA AK058135.1 - 9 - 10 1100 90
AK093455.1 - 1 - 2 1158 90
AK096525.1 - 9 - 10 2278 90
AK223343.1 - 9 - 10 1085 90
AK315123.1 - 8 - 9 1141 90
AL833828.1 - 9 - 10 1099 90
BC033732.1 - 9 - 10 1066 90
U70824.1 - 9 - 10 1124 90
U70880.1 - 8 - 9 1109 90

View Full Product Details