Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
Chromosome Location
Chr.3: 50341106 - 50346028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308379.1 NP_001295308.1 9 - 10 1273 76
NM_015896.3 NP_056980.2 10 - 11 1288 76
XM_005265216.2 XP_005265273.1 9 - 10 1461 76
GenBank mRNA AK058135.1 - 10 - 11 1218 76
AK093455.1 - 2 - 3 1276 76
AK096525.1 - 10 - 11 2396 76
AK223343.1 - 10 - 11 1203 76
AK315123.1 - 9 - 10 1259 76
AL833828.1 - 10 - 11 1217 76
BC033732.1 - 10 - 11 1184 76
U70824.1 - 10 - 11 1242 76
U70880.1 - 9 - 10 1227 76

View Full Product Details