The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288973.1 | NP_001275902.1 | 13 - 14 | 1830 | 54 |
NM_001288974.1 | NP_001275903.1 | 13 - 14 | 1830 | 54 | |
NM_001288975.1 | NP_001275904.1 | 13 - 14 | 1830 | 54 | |
NM_003474.5 | NP_003465.3 | 13 - 14 | 1839 | 54 | |
NM_021641.4 | NP_067673.2 | 13 - 14 | 1839 | 54 | |
XM_017016705.1 | XP_016872194.1 | 12 - 13 | 1742 | 54 | |
XM_017016706.1 | XP_016872195.1 | 3 - 4 | 881 | 54 | |
GenBank mRNA | AF023476.2 | - | 13 - 14 | 1732 | 54 |
AF023477.2 | - | 13 - 14 | 1732 | 54 | |
AK127856.1 | - | 13 - 14 | 1735 | 54 | |
AY358878.1 | - | 13 - 14 | 1727 | 54 | |
BC060804.1 | - | 13 - 14 | 1769 | 54 | |
KF444157.1 | - | 13 - 14 | 1830 | 54 |