The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283041.1 | NP_001269970.1 | 12 - 13 | 1667 | 128 |
NM_001283042.1 | NP_001269971.1 | 12 - 13 | 1667 | 128 | |
NM_013396.4 | NP_037528.3 | 12 - 13 | 1667 | 128 | |
XM_006723992.3 | XP_006724055.1 | 5 - 6 | 519 | 128 | |
XM_011529529.1 | XP_011527831.1 | 12 - 13 | 1745 | 128 | |
XM_011529531.2 | XP_011527833.1 | 11 - 12 | 1442 | 128 | |
XM_011529533.2 | XP_011527835.1 | 5 - 6 | 504 | 128 | |
XM_011529535.2 | XP_011527837.1 | 12 - 13 | 1745 | 128 | |
XM_017028321.1 | XP_016883810.1 | 12 - 13 | 1348 | 128 | |
XM_017028322.1 | XP_016883811.1 | 11 - 12 | 1621 | 128 | |
XM_017028323.1 | XP_016883812.1 | 10 - 11 | 1066 | 128 | |
XM_017028324.1 | XP_016883813.1 | 12 - 13 | 1519 | 128 | |
XM_017028325.1 | XP_016883814.1 | 12 - 13 | 1745 | 128 | |
XM_017028327.1 | XP_016883816.1 | 7 - 8 | 679 | 128 | |
XM_017028328.1 | XP_016883817.1 | 12 - 13 | 1745 | 128 | |
XM_017028329.1 | XP_016883818.1 | 7 - 8 | 679 | 128 | |
GenBank mRNA | AF134213.1 | - | 12 - 13 | 1497 | 128 |
AF170562.1 | - | 12 - 13 | 1667 | 128 | |
AK022574.1 | - | 3 - 4 | 350 | 128 | |
BC075792.1 | - | 12 - 13 | 1668 | 128 | |
FJ763652.1 | - | 12 - 13 | 1298 | 128 |