Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP25
Entrez Gene ID
29761
Gene Name
ubiquitin specific peptidase 25
Gene Aliases
USP21
Chromosome Location
Chr.21: 15729947 - 15880072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283041.1 NP_001269970.1 15 - 16 2124 97
NM_001283042.1 NP_001269971.1 15 - 16 2124 97
NM_013396.4 NP_037528.3 15 - 16 2124 97
XM_005260949.4 XP_005261006.1 5 - 6 673 97
XM_006723992.3 XP_006724055.1 8 - 9 976 97
XM_011529529.1 XP_011527831.1 15 - 16 2202 97
XM_011529531.2 XP_011527833.1 14 - 15 1899 97
XM_011529533.2 XP_011527835.1 8 - 9 961 97
XM_017028321.1 XP_016883810.1 15 - 16 1805 97
XM_017028322.1 XP_016883811.1 14 - 15 2078 97
XM_017028323.1 XP_016883812.1 13 - 14 1523 97
XM_017028324.1 XP_016883813.1 15 - 16 1976 97
XM_017028325.1 XP_016883814.1 15 - 16 2202 97
XM_017028327.1 XP_016883816.1 10 - 11 1136 97
XM_017028328.1 XP_016883817.1 15 - 16 2202 97
XM_017028329.1 XP_016883818.1 10 - 11 1136 97
GenBank mRNA AF134213.1 - 15 - 16 1954 97
AF170562.1 - 15 - 16 2124 97
AK022447.1 - 3 - 4 426 97
AK022574.1 - 6 - 7 807 97
BC075792.1 - 15 - 16 2125 97
FJ763652.1 - 15 - 16 1755 97
HG765100.1 - 5 - 6 657 97

View Full Product Details