Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAXD
Entrez Gene ID
55739
Gene Name
NAD(P)HX dehydratase
Gene Aliases
CARKD, LP3298
Chromosome Location
Chr.13: 110615460 - 110639996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK298917.1 - 1 - 2 114 99

View Full Product Details