Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF1
Entrez Gene ID
9138
Gene Name
Rho guanine nucleotide exchange factor 1
Gene Aliases
GEF1, LBCL2, LSC, P115-RHOGEF, SUB1.5
Chromosome Location
Chr.19: 41883200 - 41907454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198977.1 NP_945328.1 4 - 5 335 75
GenBank mRNA BC011726.2 - 4 - 5 310 75
BC067262.1 - 4 - 5 278 75

View Full Product Details